Order Entry
Mexico
Orders LinkContactUsLinkComponent
Porous Cups

Porous Cups

Porous pots, cylindrical, 25×76 mm
Catalog # 470149-184
Número de proveedor #:  J264
Porous Cups
Catalog # 470149-184
Porous Cups
Porous Cups

Porous pots, cylindrical, 25×76 mm

Price
$60.19
Log in Para disponibilidad
Each (1UN) Case de 24 (1UN)
Product Details & Documents

Porous cylindrical cups.

  • Made of ceramic

Pore diameter is approximately 0.5 µm.

Especificaciones
  • Description:
    Porous pots, cylindrical
  • Diameter ×H:
    25×76 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.