Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Precision Linear Expansion Apparatus

Rod, Linear Expansion, Copper
Catalog # 470149-176
Supplier:   VWR International
Precision Linear Expansion Apparatus
Catalog # 470149-176
Supplier:  VWR International
Precision Linear Expansion Apparatus
Precision Linear Expansion Apparatus

Rod, Linear Expansion, Copper

Price
$318.17
Log in for availability
Each (1UN) Case de 50 (1UN)
Product Details & Documents

The Precision Linear Expansion Apparatus produces accurate determinations of the coefficient of expansion.

  • For Measuring Thermal Expansion of Metals
  • Choice of Four Kinds of Rods: Aluminum, Steel, Copper, and Brass
  • No Electrical Accessories Available
  • Readings Are Repeatable to 0.005 mm
  • Easy-To-Replace Expansion Rods

This rigidly constructed apparatus requires no electrical accessories, and readings are repeatable to 0.005 mm. Expansion rods can be easily replaced without removing stoppers by sliding one rod into the heating jacket that automatically ejects the other rod.

Four expansion rods are available: aluminum, brass, copper, and steel. Rods measure 60 cm x 6.4 mm (1/4") in diameter. A steam generator is required.

Specifications
  • Description:
    Rod, Linear Expansion, Copper