Large Wave form helix
Large Wave Form Helix, No Stand
Catalog # 470148-992
Supplier: VWR International
Jump to:
Product Details & Documents
The Large Wave form helix easily demonstrates wave phenomena.
- Students Research Behavior of Pulses, Traveling Waves, and Standing Waves
- Stand Makes for Easy Storage
- For Demonstrating Wave Phenomena
With this demonstration, students can conceptualize the behavior of pulses, traveling waves, and standing waves.
With tangling woes also an issue, the featured stand provides easy deployment, retrieval, and storage.
Closed length of spring is 10″ long and 2¹/₂″ diameter. Base is 5×5″.
Especificaciones
- Description:Large Wave Form Helix, No Stand