Vinyl Lab Apron
Adult Apron, Clear, 35×45"
470148-718
Supplier #: VNAP-AD
Tough vinyl lab aprons are resistant to lab reagents, acids, alkalis, oils, and grease.
- Halter Design
- Aprons Do Not Restrict Movement
- Two Sizes Available
- Resistant to Acids, Alkalis, Oils, and More
- Reinforced Strain Points
- Latex free
The halter design keeps arms free for unrestricted movement in the lab.
The aprons are reinforced at strain points.
- Color:Clear
- Description:Adult Apron
- W×L:35×45"