Alloy Tuning Fork Sets
Tuning Forks, Set of 4
Catalog # 470148-640
Supplier: VWR International
Jump to:
Product Details & Documents
Two sets of popular tuning forks are available.
- Useful Selection of Frequencies
- Two Sets Available
- Setting from A to G
One set features four tuning forks (256 C, 320 E, 384 G, 512C).
The other set features eight tuning forks (256 C, 320 E, 384 G, 480 B, 288 D, 341.3 F, 426.7 A, 512 C).
Especificaciones
- Description:Tuning Forks, Set of 4