Order Entry
Mexico
Orders LinkContactUsLinkComponent
Square Glass Coverslips

Square Glass Coverslips

Glass cover slips, square, 25×25 mm, 0.15 mm, #1
Catalog # 470145-754
Supplier #:  94-25252
Square Glass Coverslips
Catalog # 470145-754
Square Glass Coverslips
Square Glass Coverslips

Glass cover slips, square, 25×25 mm, 0.15 mm, #1

Price
$87.42
Log in for availability
Onza
Product Details & Documents

Coverslip One is 0.15 mm thick. There are approximately 100–115 coverslips per ounce.

  • Non corrosive glass
  • Pre-cleaned

Coverslip Two is 0.2 mm thick. There are approximately 75–100 coverslips per ounce.

 

Specifications
  • Numbering:
    #1
  • Description:
    Glass cover slips, square
  • L×W:
    25×25 mm
  • Thickness:
    0.15 mm