Double Sided Mirrors
Mirrors, large, 8.5x11"
Catalog # 470144-530
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: MSA8X11-F
Jump to:
Product Details & Documents
Flexible mirrors for optical experiments.
- Acts as a concave or convex mirror
- Can be cut with scissors
You'll get lots of use out of these double-sided, flexible plastic mirrors that can be used for investigations, observations and creative activities involving reflection, symmetry and more. The mirrors can be cut with scissors.
Especificaciones
- Description:Mirrors, large, 8.5x11"