Sunprint Kit
Kit, Sunprint, 12 Sheets, 10 cm x 10 cm each
Catalog # 470106-170
Supplier: UNIVERSITY OF CALIFORNIA
Número de proveedor #: W110A
Jump to:
Product Details & Documents
The Sunprint Kit includes sun sensitive paper to help you make photographic prints using the power of the sun.
To make images, just place an array of interesting objects on a sheet of Sunprint™ paper, expose it to the sun for a few minutes, immerse in water and watch as a permanent image appears.
Delivery information: Includes 12 sheets of sun-sensitive paper (10×10cm), acrylic plate and instructions.
Especificaciones
- Description:Kit, Sunprint, 12 Sheets, 10 cm x 10 cm each
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



