Double Sided Mirrors
Mirror, Medium, 4x6"
Catalog # 470105-958
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: MSA4X6-F
Jump to:
Product Details & Documents
Flexible mirrors for optical experiments.
- Acts as a concave or convex mirror
- Can be cut with scissors
You'll get lots of use out of these double-sided, flexible plastic mirrors that can be used for investigations, observations and creative activities involving reflection, symmetry and more. The mirrors can be cut with scissors.
Especificaciones
- Description:Mirror, Medium, 4x6"