Viable Pea Seeds
Alaska Green ( Smooth) Seeds, 8 g
470105-818
: 821*1 OZ.
Learn genetics like Mendel.
- Seeds of multiple Phenotypes
- Variable sizes for each seed type
Use these peas in any lab activity, not just genetics. Peas are an excellent option for germination, growth, and genetic experiments.
: Where a Count of Seeds is indicted, the Quantity is an estimate based on Weight and may vary slightly.
- Description:Alaska Green ( Smooth) Seeds
- Size:8 g