Order Entry
Mexico
Orders LinkContactUsLinkComponent
Enameled Copper Magnet Wire

Enameled Copper Magnet Wire

Wire, Enameled Copper Magnet, 22 Gauge, 149 m, 454 grams
Catalog # 470104-612
Número de proveedor #:  E-22
Enameled Copper Magnet Wire
Catalog # 470104-612
Enameled Copper Magnet Wire

Wire, Enameled Copper Magnet, 22 Gauge, 149 m, 454 grams

Price
$763.26
Log in Para disponibilidad
Each (1UN)
Product Details & Documents

A high quality enameled, copper magnet wire.

The 454 g spools are available in several standard AWG sizes.

Especificaciones
  • Description:
    Wire, Enameled Copper Magnet, 22 Gauge, 149 m, 454 grams
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.