Wooden Splints
Wooden Splints, 114 mm, 4 mm
Catalog # 470104-504
Supplier: PRECISION LABORATORIES SE
Número de proveedor #: WDS-1B-100
Jump to:
Product Details & Documents
Wooden splints are used for mixing and applying in a laboratory setting.
Especificaciones
- Description:Wooden Splints
- Height:114 mm
- Width:4 mm