Pliers
Pliers, Forged steel, 61/2", Long Nose
Catalog # 470104-436
Supplier: GREAT NECK TOOLS, INC.
Número de proveedor #: LN65C
Jump to:
Product Details & Documents
These pliers, available in various styles, are made of drop forged steel.
- Heat treated with fully polished heads and vinyl grips
Certifications: All meet ANSI tool standards and are covered by a lifetime warranty.
Especificaciones
- Description:Pliers
- Material:Forged steel
- Size:61/2"
- Tip Style:Long Nose