Scissors—Superior
Straight Surgical Scissors—Superior
Catalog # 470104-198
Supplier: DUNRITE INSTRUMENTS SE
Supplier #: 6157000
Jump to:
Product Details & Documents
Our finest scissors.
- Machine cut edges for a consistent cut
- Brushed stainless finish
- Stainless construction resists rust
Our best grade dissecting scissors feature fine sharp points and machined cut edges. Open shank. Brushed stainless steel.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
- Description:Straight Surgical Scissors—Superior