Orders LinkContactUsLinkComponent
Polyethylene Dispensing Bottles
Polyethylene Dispensing Bottles
Bottle, Dispensing, 118 mL
Catalog # 470100-720
Número de proveedor #:  395 & 93
Polyethylene Dispensing Bottles
Catalog # 470100-720
Polyethylene Dispensing Bottles
Polyethylene Dispensing Bottles
Bottle, Dispensing, 118 mL
Price
$368.83
Log in Para disponibilidad
Paquete de 12
Product Details & Documents
Low-density polyethylene bottles and spouts with sealer caps that snap on securely. Spout openings are standard 0.030" and can be enlarged with scissors.
Especificaciones
  • Description:
    Bottle, Dispensing, 118 mL