Polyethylene Dispensing Bottles
Bottle, Dispensing, 118 mL
Catalog # 470100-720
Supplier: PLASTIC BOTTLES INC.
Número de proveedor #: 395 & 93
Jump to:
Product Details & Documents
Low-density polyethylene bottles and spouts with sealer caps that snap on securely. Spout openings are standard 0.030" and can be enlarged with scissors.
Especificaciones
- Description:Bottle, Dispensing, 118 mL