Orders LinkContactUsLinkComponent
Polyethylene Dispensing Bottles
Bottle, Dispensing, 30 mL
  470100-718
 :  300 & 351
Polyethylene Dispensing Bottles
  470100-718
Polyethylene Dispensing Bottles
Polyethylene Dispensing Bottles
Bottle, Dispensing, 30 mL
$256.91
Paquete de 12
Low-density polyethylene bottles and spouts with sealer caps that snap on securely. Spout openings are standard 0.030" and can be enlarged with scissors.
  • Description:
    Bottle, Dispensing, 30 mL