Orders LinkContactUsLinkComponent
BoneClones® Replica Claws
BoneClones® Replica Claws
Polar bear claw, 1, 9 cm
Catalog # 470095-130
Supplier #:  KO-048
BoneClones® Replica Claws
Catalog # 470095-130
BoneClones® Replica Claws
BoneClones® Replica Claws
Polar bear claw, 1, 9 cm
Price
$652.93
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Compare the Claws of a Carnivore and a Herbivore.
  • Explore the influences of habitat and use in claw shape
  • Highly detailed replicas from real specimens
In the animal world claws aren't just for hunting. Based on what your students can research about the animals, can they explain the morphological differences and similarities between these claws? Each claw is a replica cast from a real specimen.
Especificaciones
  • Description:
    Polar bear claw
  • Length:
    9 cm
  • No. of Parts:
    1