Order Entry
Mexico
Orders LinkContactUsLinkComponent
Tape Measure

Tape Measure

Lead-free tape measure, metric and imperial, 1.5 m (5 ft.)
Catalog # 470094-020
Número de proveedor #:  2991377
Tape Measure
Catalog # 470094-020
Tape Measure
Tape Measure

Lead-free tape measure, metric and imperial, 1.5 m (5 ft.)

Price
$21.60
Log in Para disponibilidad
Each (1UN) Caja de 250 (1UN)
Product Details & Documents

1.5m dual scale (inches and centimeters) tape measures.

  • Dual scale
  • Plastic with metal tabs

Measures are plastic with metal tabs at the ends.

Especificaciones
  • Description:
    Lead-free tape measure, metric and imperial
  • Length:
    1.5 m (5 ft.)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.