$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
BRACHIOPODREPL FOSSIL CAST 3/4X3/4 RES/A
470093-892
:
SKYWILL FAR EAST INDUSTRIES
BRACHIOPODREPL FOSSIL CAST 3/4X3/4 RES/A
470093-892
:
SKYWILL FAR EAST INDUSTRIES
:
2991280
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
: VWR International