Straws, Plastic
Straws, Wrapped, 0.58 cm, 19.7 cm, White
Catalog # 470092-512
Supplier: Wards
Número de proveedor #: 1500 WHITE
Jump to:
Product Details & Documents
Straws made of plastic.
Especificaciones
- Color:White
- Description:Straws, Wrapped
- Length:19.7 cm
- Diameter:0.58 cm