Histology Prepared Microscope Slides
Muscle, Smooth (Intestine), Cross Section, Longitudinal Section, Hematoxylin and Eosin, Animal, Canis lupis familiaris
Catalog # 470089-482
Supplier: C & A SCIENTIFIC CO INC
Número de proveedor #: 4H-55
Jump to:
Product Details & Documents
Great for beginners.
- Professionally stained samples for good visualization
- Perfect for your introductory biology classrooms
Stock-up on these affordable prepared microscope slides for your classroom. These slides are a great way to introduce your students to viewing samples under a microscope.
Delivery information: These slides are made with care, by our manufacturing partners worldwide.
Especificaciones
- Type:Animal, Canis lupis familiaris
- Description:Muscle, Smooth (Intestine)
- Stain:Hematoxylin and Eosin
- Orientation:Cross Section, Longitudinal Section