Order Entry
Mexico
Orders LinkContactUsLinkComponent
Scheibler Desiccator

Scheibler Desiccator

Desiccator Plate, —, 190 mm, 30 mm, 7
Catalog # 470174-666
Scheibler Desiccator
Catalog # 470174-666
Supplier:  
Scheibler Desiccator
Scheibler Desiccator

Desiccator Plate, —, 190 mm, 30 mm, 7

Discontinued
Product Details & Documents

Non-Vacuum Desiccator

  • Made of heavy, flint glass

Made of clear glass with thick uniform walls and a heavy, broad flange. Cover with molded knob is ground to fit the flange perfectly.

Specifications
  • Hole Ø:
    30 mm
  • Ø int.:
  • No. of Holes:
    7
  • Description:
    Desiccator Plate
  • Plate Diameter:
    190 mm