Tri Pour Beakers
400 ml (14 oz.), Tri-pour beakers
Catalog # 470174-582
Supplier #: 80096
Jump to:
Discontinued
El producto ha sido descontinuado. Sin embargo, encontramos productos alternativos similares.
Alternativas
VWR BEAKER TRIPOUR 400ML CS100 Catalog #25384-156
Product 25384-156 is similar to 470174-582 but manufacturer and specifications may vary.
Product Details & Documents
These unbreakable polypropylene beakers feature a tri-cornered lip for dripless pouring.
Accurate graduations to ±5
- Tri-cornered lip for dripless pouring
- Unbreakable
- Resistant to common acids, solvents, and alkalis
- Autoclavable
They are unaffected by commonly used acids, alkalis, and solvents and are autoclavable.
Graduations are accurate to ±5.
Especificaciones
- Capacity:400 ml (14 oz.)
- Description:Tri-pour beakers