Y-Shaped Connectors
Polypropylene Y Connectors, Fits 3/16" Tubing
Catalog # 470174-272
Supplier: DYNALON
Número de proveedor #: 222555
Jump to:
Product Details & Documents
Divide single source tubing into two with this autoclavable connector. Fine chemically resistant polypropylene. Package of ten.
Especificaciones
- Description:Polypropylene Y Connectors, Fits 3/16" Tubing