Orders LinkContactUsLinkComponent
Y-Shaped Connectors
Y-Shaped Connectors
Polypropylene Y Connectors, Fits 3/16" Tubing
Catalog # 470174-272
Supplier:   DYNALON
Número de proveedor #:  222555
Y-Shaped Connectors
Catalog # 470174-272
Supplier:  DYNALON
Y-Shaped Connectors
Y-Shaped Connectors
Polypropylene Y Connectors, Fits 3/16" Tubing
Price
$525.07
Log in Para disponibilidad
Paquete de 10 Case de 10 (10UN)
Product Details & Documents
Divide single source tubing into two with this autoclavable connector. Fine chemically resistant polypropylene. Package of ten.
Especificaciones
  • Description:
    Polypropylene Y Connectors, Fits 3/16" Tubing