Orders LinkContactUsLinkComponent
Single Factor Water Test Kits
Single Factor Water Test Kits
Single factor water test kit, total chlorine, 30 tests
Catalog # 470173-832
Número de proveedor #:  481110
Single Factor Water Test Kits
Catalog # 470173-832
Single Factor Water Test Kits
Single Factor Water Test Kits
Single factor water test kit, total chlorine, 30 tests
Price
$456.84
Log in Para disponibilidad
Each (1UN) Case de 12 (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Simply Dip in Water and Read the Results
These kits feature a wide range of test factors for a fraction of the price of other single factor test kits. These test kits also eliminate the need for liquid reagents, hazardous chemicals, and glassware. Each kit comes with instructions.
Especificaciones
  • Description:
    Single factor water test kit, total chlorine, 30 tests
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.