Orders LinkContactUsLinkComponent
Magnetic Stirring Head
Magnetic Stirring Head
Magnetic Stirring Head
Catalog # 470161-312
Supplier:   DYNALON
Número de proveedor #:  304455-0003
Magnetic Stirring Head
Catalog # 470161-312
Supplier:  DYNALON
Magnetic Stirring Head
Magnetic Stirring Head
Magnetic Stirring Head
Price
$406.96
Log in Para disponibilidad
Each (1UN) Case de 12 (1UN)
Product Details & Documents
The paddle configuration is quiet, with large vortex for thorough mixing.
Lower rpms make rods suitable for air or water-driven mixing units. Size: 1×1".
Especificaciones
  • Description:
    Magnetic Stirring Head