Fast Release Pi-Pumps
Fast Release Pi-Pump, 2 ml, Blue
470153-806
Supplier #: F37904-0002
Designed for fast and accurate one-handed operation, this pump will reduce the risk of injury from repetitive pipetting.
- Each pump is resistant to acids, bases, and most solvents
- Disassembly for cleaning is easy
The fast-release trigger expels the entire contents at one shot, or you can use the thumb wheel for precision dispensing.
- Capacity:2 ml
- Color:Blue
- Description:Fast Release Pi-Pump