Plane Mirrors
Glass mirrors with seamed edges,152x100x6.3 mm
Catalog # 470153-012
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: MSW6X4-PK/6
Jump to:
Product Details & Documents
All Types and Sizes for All Optical Applications
- Front surface silvering eliminates double reflections
- All produce virtual images
These plane mirrors are rectangular. They are available in glass or shatterproof Plexiglas. Back surface silvered mirrors are less expensive, and ideal for general use. The silvered back surface is painted for durability. Front surface silvering eliminates double reflections at the unsilvered surface, giving a brighter, sharper image. However, these mirrors are more easily damaged by scratching.
Especificaciones
- Description:Glass mirrors with seamed edges,152x100x6.3 mm