Orders LinkContactUsLinkComponent
Cylinder Guards
Cylinder Guards, For Cylinder Cap., 5 – 10 mL
  470151-208
 :  VCYCG1
Cylinder Guards
  470151-208
 :  
Cylinder Guards
Cylinder Guards
Cylinder Guards, For Cylinder Cap., 5 – 10 mL
Black sponge rubber guards prevent breakage and rolling.
  • Hexagonal
  • 13 mm thick
  • Description:
    Cylinder Guards, For Cylinder Cap., 5 – 10 mL