Orders LinkContactUsLinkComponent
Collection Boxes
Collection Boxes, 9 cell
  470151-142
 :  0145-001
Collection Boxes
  470151-142
Collection Boxes
Collection Boxes
Collection Boxes, 9 cell
$159.92
Each (1UN)
Create your own collections with these study boxes. Complete with styrene inserts and lids. Choose how many compartments you want.
  • Three box sizes available
WL7075L-09
9 Cell Box
Box: 7 x 4.75 inches
Cells: 1.25 x 2 inches
WL7075L-15
15 Cell Box
Box: 11 x 7 inches
Cells: 1.75 x 1.75 inches
WL7075-24
24 Cell Box
Box: 11 x 7 inches
Cells: 1.5 x 1.25 inches
  • Description:
    Collection Boxes, 9 cell