Orders LinkContactUsLinkComponent
Square Glass Coverslips
  470151-078
Square Glass Coverslips
  470151-078
 :  
Square Glass Coverslips
Square Glass Coverslips
Coverslip One is 0.15 mm thick. There are approximately 100–115 coverslips per ounce.
  • Non corrosive glass
  • Pre-cleaned
Coverslip Two is 0.2 mm thick. There are approximately 75–100 coverslips per ounce.