Coverslip One is 0.15 mm thick. There are approximately 100–115 coverslips per ounce.
Coverslip Two is 0.2 mm thick. There are approximately 75–100 coverslips per ounce.
#: 16001-188
: VWR International
#: 76322-154
#: 82003-820
#: 76518-336
#: 25384-342
#: 75816-100
#: 47729-576
#: 89428-752
#: 89107-770