Square Glass Coverslips
Catalog # 470151-074
Discontinued
El producto ha sido descontinuado. Sin embargo, encontramos productos alternativos similares.
Alternativas
COVER GLASS 2 THICK18 MM BOREAL 1 OZ Catalog #470145-878
El producto 470145-878 es similar a 470151-074, pero el fabricante y las especificaciones pueden variar.
Product Details & Documents
Coverslip One is 0.15 mm thick. There are approximately 100–115 coverslips per ounce.
- Non corrosive glass
- Pre-cleaned
Coverslip Two is 0.2 mm thick. There are approximately 75–100 coverslips per ounce.
Documents
Certificates
Buscar por número de lote Recommendations