Square Glass Coverslips
Glass cover slips, square, 18×18 mm, 0.20 mm, #2
Catalog # 470151-072
Jump to:
Discontinued
El producto ha sido descontinuado. Sin embargo, encontramos productos alternativos similares.
Alternativas
Product Details & Documents
Coverslip One is 0.15 mm thick. There are approximately 100–115 coverslips per ounce.
- Non corrosive glass
- Pre-cleaned
Coverslip Two is 0.2 mm thick. There are approximately 75–100 coverslips per ounce.
Certificates
Buscar por número de loteEspecificaciones
- Numbering:#2
- Description:Glass cover slips, square
- L×W:18×18 mm
- Thickness:0.20 mm



