Orders LinkContactUsLinkComponent
United Scientific™ Push/Pull Spring Scales
United Scientific™ Push/Pull Spring Scales
Push/pull spring scale, cream, 3000 g/30 N, graduation interval: 0.5 N
Catalog # 470150-868
Número de proveedor #:  PS3000
United Scientific™ Push/Pull Spring Scales
Catalog # 470150-868
United Scientific™ Push/Pull Spring Scales
United Scientific™ Push/Pull Spring Scales
Push/pull spring scale, cream, 3000 g/30 N, graduation interval: 0.5 N
Price
$140.20
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Versatile spring scale.
  • Weighs in Grams and Newtons
  • Color coded to differentiate capacity
These Push Pull Spring Scales are a versatile spring scale that can be used to measure forces using push or pull functions, or to weigh a suspended object from the large hook.
Delivery information: Includes a zero adjustment screw, a metal suspension ring, instructions and application sheets.
Caution: This product is for ages six and up.
Especificaciones
  • Description:
    Push/pull spring scale, cream
  • Weighing Capacity:
    3000 g/30 N, graduation interval: 0.5 N