Nesting Plant Trays
Nesting Plant Trays, 23"L x 12"W x 2 1/2"H
Catalog # 470150-492
Supplier: GROWERS SUPPLY CO.
Supplier #: GS2211-B
Jump to:
Product Details & Documents
Perfect for starting seeds, the trays are made of durable plastic, so they won't rot or harbor bacteria like wooden flats.
- They are watertight, washable, and stack neatly to store
Ordering information: Available in tan color only.
Specifications
- Description:Nesting Plant Trays, 23"L x 12"W x 2 1/2"H