Orders LinkContactUsLinkComponent
Cartridge-Type Deionizers
Universal cartridge, 1600 grains (as CaCO3) ion exchange capacity, 20½", 27 L/hr
  470150-334
 :  1X-02-4040
Cartridge-Type Deionizers
  470150-334
Cartridge-Type Deionizers
Cartridge-Type Deionizers
Universal cartridge, 1600 grains (as CaCO3) ion exchange capacity, 20½", 27 L/hr
$1,591.47
Each (1UN) Case de 6 (1UN)
Great for lab use.
  • Research model cartridge removes essentially all ionizable constituents including silica and free carbon dioxide
  • Universal model cartridge; does not remove silica or carbon dioxide
  • Single cartridge holder with faucet adapter (includes a brass control valve, inlet hose and outlet tubing), consists of one heavy-duty stamped steel mounting fixture, pressure fitting, tubing and washers
Transparent, plastic cartridges contain ion-exchange resins of the mixed-bed type for providing high purity water at low flow rates. For purification either by intermittent or steady flow. Intended for wall mounting, they are designed for operation at pressures up to 100 psi with a max flow of 27 liters per hour in the single cartridge holder.
  • Flow rate:
    27 L/hr
  • Description:
    Universal cartridge, 1600 grains (as CaCO3) ion exchange capacity
  • Length:
    20½"