Orders LinkContactUsLinkComponent
VWR® Standard Standard-Grade Erlenmeyer Flasks
VWR® Standard Standard-Grade Erlenmeyer Flasks
25 ml, 10 - 25 ml, 0
Catalog # 470149-316
Supplier:   VWR International
VWR® Standard Standard-Grade Erlenmeyer Flasks
Catalog # 470149-316
Supplier:  VWR International
VWR® Standard Standard-Grade Erlenmeyer Flasks
VWR® Standard Standard-Grade Erlenmeyer Flasks
25 ml, 10 - 25 ml, 0
Price
$589.58
Log in Para disponibilidad
Paquete de 12 Case de 4 (12UN)
Product Details & Documents
Our high-quality borosilicate standard-grade Erlenmeyer flasks meet ASTM standards at a great value.
  • Uniformly thick walls
  • White enamel graduated markings
  • High-quality borosilicate glass construction
With thick, uniform wall thickness; white enamel metric scale; and a marking spot, each flask can be heated and used with acids, salt solutions, and organic solvents.
Documents
Especificaciones
  • Graduation Range:
    10 - 25 ml
  • Capacity:
    25 ml
  • Stopper No.:
    0