VWR® Standard Standard-Grade Erlenmeyer Flasks
500 ml, 100 - 500 ml, 7
Supplier: VWR International
Jump to:
Product Details & Documents
Our high-quality borosilicate standard-grade Erlenmeyer flasks meet ASTM standards at a great value.
- Uniformly thick walls
- White enamel graduated markings
- High-quality borosilicate glass construction
With thick, uniform wall thickness; white enamel metric scale; and a marking spot, each flask can be heated and used with acids, salt solutions, and organic solvents.
Documents
Certificates
Search by Lot NumberSpecifications
- Graduation Range:100 - 500 ml
- Capacity:500 ml
- Stopper No.:7