Orders LinkContactUsLinkComponent
VWR® Standard Standard-Grade Erlenmeyer Flasks
VWR® Standard Standard-Grade Erlenmeyer Flasks
500 ml, 100 - 500 ml, 7
Catalog # 470149-308
Supplier:   VWR International
VWR® Standard Standard-Grade Erlenmeyer Flasks
Catalog # 470149-308
Supplier:  VWR International
VWR® Standard Standard-Grade Erlenmeyer Flasks
VWR® Standard Standard-Grade Erlenmeyer Flasks
500 ml, 100 - 500 ml, 7
Price
$883.03
Log in for availability
Paquete de 6 Case de 6 (6UN)
Product Details & Documents
Our high-quality borosilicate standard-grade Erlenmeyer flasks meet ASTM standards at a great value.
  • Uniformly thick walls
  • White enamel graduated markings
  • High-quality borosilicate glass construction
With thick, uniform wall thickness; white enamel metric scale; and a marking spot, each flask can be heated and used with acids, salt solutions, and organic solvents.
Documents
Specifications
  • Graduation Range:
    100 - 500 ml
  • Capacity:
    500 ml
  • Stopper No.:
    7