VWR® Florence Boiling Flasks
500 ml, 6
Supplier: VWR International
Jump to:
Product Details & Documents
High Quality Borosilicate Glass
- Meet ASTM standards
- Round body and long neck are ideal for boiling liquids without excessively rapid evaporation
Ordering information: Rubber stoppers are not included.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Capacity:500 ml
- Rubber Stopper Size:6