VWR® Test Tubes
3 ml, —, 10×75 mm
Supplier: VWR International
Jump to:
Product Details & Documents
VWR® Test tubes are made from high-quality glass.
- Borosilicate glass construction
- Uniform thickness
Each tube features a strong lip.
Certifications: ASTM standards.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Capacity:3 ml
- Stopper Size:—
- O.D.×L:10×75 mm