VWR® Test Tubes
20 ml, 00, 16×150 mm
Supplier: VWR International
Jump to:
Product Details & Documents
VWR® Test tubes are made from high-quality glass.
- Borosilicate glass construction
- Uniform thickness
Each tube features a strong lip.
Certifications: ASTM standards.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Capacity:20 ml
- Stopper Size:00
- O.D.×L:16×150 mm