VWR® Test Tubes
55 ml, 4, 25×150 mm
Supplier: VWR International
Jump to:
Product Details & Documents
VWR® Test tubes are made from high-quality glass.
- Borosilicate glass construction
- Uniform thickness
Each tube features a strong lip.
Certifications: ASTM standards.
Documents
Certificates
Search by Lot NumberSpecifications
- Capacity:55 ml
- Stopper Size:4
- O.D.×L:25×150 mm