Order Entry
Mexico
Orders LinkContactUsLinkComponent
Test Tube Brushes

Test Tube Brushes

Brush for tube diameter 20 - 27 mm, Nylon, Tan, 29 mm
Catalog # 470149-234
Supplier:   VWR International
Test Tube Brushes
Catalog # 470149-234
Supplier:  VWR International
Test Tube Brushes
Test Tube Brushes

Brush for tube diameter 20 - 27 mm, Nylon, Tan, 29 mm

Price
$18.29
Log in Para disponibilidad
Each (1UN) Paquete de 12 (1UN)
Product Details & Documents

These test tube brushes feature a radially tufted tip.

  • Heavy, Galvanized Wire
  • Radially Tufted Tip
  • Nylon or Natural Brushes Available

They also feature heavy, galvanized wire.

Nylon or natural bristles are available.

Especificaciones
  • Bristle Color:
    Tan
  • Bristle Material:
    Nylon
  • Brush head Ø:
    29 mm
  • Description:
    Brush for tube diameter 20 - 27 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.