Bare Nichrome Wire
Bare Nichrome wire, 24 SWG
Catalog # 470149-224
Supplier: VWR International
Jump to:
Product Details & Documents
Create High Performance Loops
125gm reels of bare Nichrome wire for use in resistance heating or inoculating loops.
Specifications
- Description:Bare Nichrome wire
- Gauge:24 SWG