Order Entry
Mexico
Orders LinkContactUsLinkComponent
Black Rubber Bib Apron

Black Rubber Bib Apron

Black, M, 36×27"
Catalog # 470149-220
Número de proveedor #:  RBAP-MD
Black Rubber Bib Apron
Catalog # 470149-220
Black Rubber Bib Apron
Black Rubber Bib Apron

Black, M, 36×27"

Price
$142.96
Log in Para disponibilidad
Each (1UN) Case de 25 (1UN)
Product Details & Documents

Supply Your Lab with Personal Protective Equipment

  • Basic black rubber apron
  • Perfect for labs, kitchens, or shops
  • Contains latex

Suitable for general lab work. Comfortable and flexible Black Rubber Bib Aprons are waterproof and resistant to most chemicals used by students in the lab.

Especificaciones
  • Clothing Size:
    M
  • Color:
    Black
  • L×W:
    36×27"
  • Cat. No.:
    470149-220
  • Garment application:
    Food|Laboratory
  • Gender:
    Unisex
  • Fabric:
    Rubber