Orders LinkContactUsLinkComponent
Porcelain Buchner Funnels
2A, —, 7 cm
  470149-206
Porcelain Buchner Funnels
  470149-206
Porcelain Buchner Funnels
Porcelain Buchner Funnels
2A, —, 7 cm
$309.04
Each (1UN) Case de 6 (1UN)
High Quality yet Affordable Buchner Funnels.
  • Two sizes available
  • Fragile
These high quality Buchner funnels are made from glazed perforated plate high-quality porcelain. They have a fixed, perforated plate and are glazed on the inside and outside with exception of the rim.
 : Fragile, use care when handling.
  • Ø:
  • Size:
    2A
  • Fits Paper Diameter:
    7 cm