Orders LinkContactUsLinkComponent
VWR® Volumetric Flasks
VWR® Volumetric Flasks
2000 ml
Catalog # 470149-060
Supplier:   VWR International
VWR® Volumetric Flasks
Catalog # 470149-060
Supplier:  VWR International
VWR® Volumetric Flasks
VWR® Volumetric Flasks
2000 ml
Price
$626.14
Log in Para disponibilidad
Each (1UN) Case de 4 (1UN)
Product Details & Documents
Our borosilicate flasks meet ASTM standards.
  • Class A Flasks
  • Borosilicate Glass Construction
  • Graduated Marking for Measurements
  • Economical
  • Snap-Caps Included
The necks fit the included snap-caps.
The Class A, volumetric flasks include a graduation line for accurate measurements.
Documents
Especificaciones
  • Capacity:
    2000 ml