VWR® Linear Spring Scales
5000 g x 100 g (50 N x 5 N)
Catalog # 470149-050
Jump to:
Discontinued
El producto ha sido descontinuado. Sin embargo, encontramos productos alternativos similares.
Alternativas
Product Details & Documents
Designed for Heavy Duty Student Use
- Feature easy to read markings
- Durable for years of regular student use
- Length: 14 cm
- Each scale is easily zeroed
Each has a plated steel suspension ring at the top, and a hook at the bottom.
Especificaciones
- Weighing Capacity:5000 g x 100 g (50 N x 5 N)