Lil'pet Micropipette
250 µl
470148-996
Lil'pet Micropipettes feature ideal volumes for gel loading.
- Units accept standard micropipette tips
- Economical
- Durable
- Autoclavable
These fixed-volume pipettes feature a double stop plunger to expel all liquid from the tip.
The 1000 μl model includes two tips. All other pipettes come with one each.
- Volume:250 µl