Orders LinkContactUsLinkComponent
Lil'pet Micropipette
250 µl
  470148-996
Lil'pet Micropipette
  470148-996
Lil'pet Micropipette
Lil'pet Micropipette
250 µl
$345.18
Each (1UN) Case de 100 (1UN)
Lil'pet Micropipettes feature ideal volumes for gel loading.
  • Units accept standard micropipette tips
  • Economical
  • Durable
  • Autoclavable
These fixed-volume pipettes feature a double stop plunger to expel all liquid from the tip.
The 1000 μl model includes two tips. All other pipettes come with one each.
  • Volume:
    250 µl