Orders LinkContactUsLinkComponent
Squibb Urinometer and Jar
Squibb Urinometer and Jar
Squibb Urinometer-Hydrometer and Jar
Catalog # 470148-982
Supplier:   VWR International
Squibb Urinometer and Jar
Catalog # 470148-982
Supplier:  VWR International
Squibb Urinometer and Jar
Squibb Urinometer and Jar
Squibb Urinometer-Hydrometer and Jar
Price
$243.46
Log in Para disponibilidad
Each (1UN) Case de 20 (1UN)
Product Details & Documents
Perfect for when only minimal volumes are required, this urinometer provides highly accurate samples.
  • Calibrated, Easy-to-Read Scale
  • Range: 1.000–1.060 Sp. Gr
  • Accurate Samples
  • Stain-Free Clear Glass
  • Capacity: 50 mL
This stain-free and clear glass apparatus has a capacity of 50 mL and a wide base that prevents it from tipping.
Its range is 1.000–1.060 Sp. Gr in 0.002 graduations.
Especificaciones
  • Description:
    Squibb Urinometer-Hydrometer and Jar